Vincristine sulfate

Supplier: ADIPOGEN CORP MS

AG-CN2-0446-M005 AG-CN2-0446-M025
102988-292EA 55.38 USD
102988-292 102988-294
Vincristine sulfate
Vincristine sulphate

Immunosuppressant. Anticancer compound. Chemotherapeutic agent. Depolymerizes microtubules and irreversibly blocks binding of tubulin to microtubule proteins. Apoptosis inducer and cell cycle inhibitor in G2/M phase. Autophagy inhibitor. Inhibits microtubule-mediated fusion of autophagosome with lysosme. Inducer of IL-1beta production. Activates alone and synergistically together with doxorubicin the NLRP3-inflammasome mediated processing of IL-1beta. Monoamine oxidase (MAO) inhibitor.

Formula: C₄₆H₅₈N₄O₁₄S
MDL Number: MFCD00084729
CAS Number: 2068-78-2
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR