Betulinic acid

Supplier: ADIPOGEN CORP MS

Synonyms: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

AG-CN2-0415-M005 AG-CN2-0415-M025 AG-CN2-0415-M100
102988-188EA 41.54 USD
102988-188 102988-190 102988-192
Betulinic acid
Betulinic acid

Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.

Formula: C₃₀H₄₈O₃
MDL Number: MFCD00009619
CAS Number: 472-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR