Viomellein ≥96%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0359-C500 BVT-0359-M001
102513-792EA 180 USD
102513-792 102513-794
Viomellein ≥96%
Viomellein

Genotoxic mycotoxin. Antibacterial (Gram-positive) and insecticidal. Aromatase inhibitor.

Formula: C₃₀H₂₄O₁₁
Storage Temperature: Freezer
CAS Number: 55625-78-0

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus sulphureus.
Purity 96% (HPLC)
Appearance/Color Brown solid.
Solubility Soluble in chloroform, dichloromethane or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR