Elasnin ≥98%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0342-M001 BVT-0342-M005
102513-782EA 180 USD
102513-782 102513-784
Elasnin ≥98%
Elasnin

Inhibitor of human leukocyte elastase (IC50 = 1.3 µg/ml), different other elastases and chymotrypsin (IC50 = 82 µg/ml).

Formula: C₂₄H₄₀O₄
Storage Temperature: Freezer
CAS Number: 68112-21-0

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces sp.
Purity 98% (HPLC)
Appearance/Color Tan to brown viscous liquid.
Solubility Soluble in DMSO (5 mg/ml), methanol (5 mg/ml) or methylene chloride.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR