Papyracillic acid A (from Lachnum papyraceum/Microsphaeropsis sp) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0308-C500 BVT-0308-M001
102513-750EA 193.85 USD
102513-750 102513-752
Papyracillic acid A (from Lachnum papyraceum/Microsphaeropsis sp) ≥98% (by HPLC)
Papyracillic acid A

Papyracillic acid A is an antibiotic.


  • Off-white to beige solid


Antibiotic. Exists in an equilibrium of diastereomers.


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in dichloromethane or chloroform

Storage Temperature: Freezer
CAS Number: 960148-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR