Becatecarin (semi-synthetic) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0258-C250 BVT-0258-M001
102513-670EA 235.38 USD
102513-670 102513-672
Becatecarin (semi-synthetic) ≥98% (by HPLC)
Becatecarin

Becatecarin is a DNA intercalating agent.


  • Antibiotic
  • Topoisomerase II (Topo II) inhibitor
  • Anti-tumor compound


Semisynthetic water-soluble derivative of rebeccamycin.


Yellow solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or water (pH dependent).

Storage Temperature: Freezer
CAS Number: 119673-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR