Palmarumycin C3 ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
BVT-0078-M001
BVT-0078-M005
102513-508EA
180
USD
102513-508
102513-510
Palmarumycin C3 ≥97% (by HPLC)
Palmarumycin C3
Rasfarnesyltransferase inhibitor. Antifungal, antibacterial and herbicidal. Structurally unique natural products. The basic structure of palmarumycin C1 can be modified by a number of hydroxylation, oxygenation, dehydrogenation and chlorination steps. Antioxidant
Formula:
C₂₀H₁₂O₆ MW: 348.3 g/mol Storage Temperature: Freezer |
CAS Number:
159934-11-9 UN: 3462 ADR: 6.1,III |
Specification Test Results
Source/Host | Isolated from Sphaeropsidales sp. |
Purity |
|
Appearance/Color | Yellow to green solid. |
Solubility | Soluble in DMSO, dichloromethane, acetone or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...