Bafilomycin B1 ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

BVT-0004-C100 BVT-0004-M001 BVT-0004-M005
102513-436EA 69.23 USD
102513-436 102513-438 102513-440
Bafilomycin B1 ≥97% (by HPLC)
Bafilomycin B1

Macrolide antibiotic. Specific vacuolar-type H+-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antitrypanosomal and antileishmanial compound. Phytotoxic. Influences the cell wall biosynthesis of Asp. niger.

Formula: C₄₄H₆₅NO₁₃
Storage Temperature: Freezer
MDL Number: MFCD00242671
CAS Number: 88899-56-3
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces hygroscopicus.
Purity >97% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO or acetone. Insoluble in water. Unstable in methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR