Pseurotin A ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0003-M001 BVT-0003-M005
102513-432EA 124.62 USD
102513-432 102513-434
Pseurotin A ≥98% (by HPLC)
Pseurotin A

Pseurotin A is an antibiotic which shows nematicidal activity.


  • Cytotoxic
  • Synthase inhibitor


Antibiotic. Chitin synthase inhibitor. Cytotoxic. Shows nematicidal activity.


White to off-white powder


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in methanol or ethyl acetate; poorly soluble in water.

Formula: C₂₂H₂₅NO₈
Storage Temperature: Freezer
CAS Number: 58523-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR