6-ECDCA ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: Obeticholic acid, (R)-4-((3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, INT-747

AG-CR1-3560-M005 AG-CR1-3560-M025
102516-272EA 69.23 USD
102516-272 102516-274
6-ECDCA ≥95%
6-ECDCA

Potent and selective FXR agonist (EC50= 99nM). Induces SHP in HSCs to suppress TIMP-1 expression. Apoptosis inducer. Protects against liver fibrosis development in rat in vivo. Displays anticholeretic activity in rat in vivo. Promotes preadipocyte differentiation. Regulates adipogenesis and insulin signaling in vivo. Inhibits vascular smooth muscle cell inflammation and migration.

Formula: C₂₆H₄₄O₄
Storage Temperature: Freezer
MDL Number: MFCD16621104
CAS Number: 459789-99-2

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color White solid.
Solubility Soluble in ethanol, methanol, or DMSO. Slightly soluble in water, dichloromethane and acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR