Ebselen

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

AG-CR1-0031-M001 AG-CR1-0031-M005 AG-CR1-0031-M025
102515-994EA 20.77 USD
102515-994 102515-996 102515-998
Ebselen
Ebselen

Glutathione peroxidase mimetic. Peroxynitrite scavenger. Anti-inflammatory Antioxidant. Protein kinase C (PKC), NADPH, lipoxygenase, COX, NOS, H+-ATPase and NADPH oxidase inhibitor. Antifungal. Review. Potent IDO-1 inhibitor with Ki 94nM. Inhibitor of HIV-1 capsid C-terminal domain dimerization.

Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: 6.1 III

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR