Latrunculin A ≥96%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one

AG-CN2-0027-C100 AG-CN2-0027-C500
102515-768EA 173.08 USD
102515-768 102515-770
Latrunculin A ≥96%
Latrunculin A

Cell permeable marine toxin. Disrupts microfilament-mediated processes. Actin polymerization inhibitor in vitro and in vivo by the formation of a 1:1 complex with monomeric G-actin. Depolymerizes actin filaments (F-actin). Potent phagocytosis inhibitor. Anticancer compound. Suppresses hypoxia-induced HIF-1 activation in tumor cells. Inhibits tumor cell invasion. Acts via a different mechanism than cytochalasins.

Formula: C₂₂H₃₁NO₅S
Storage Temperature: Freezer
CAS Number: 76343-93-6

Order Now

Specification Test Results

Source/Host Isolated from Latrunculia magnifica.
Purity 96% (HPLC)
Appearance/Color White to light yellow solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR