Zerumbone ≥99%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (2E,6E,10E)-2,6,9,9-Tetramethylcycloundeca-2,6,10-trienone

AG-CN2-0004-M010 AG-CN2-0004-M050
102515-668EA 76.15 USD
102515-668 102515-670
Zerumbone ≥99%
Zerumbone

Chemopreventive probe. Apoptosis inducer. Potent anti-tumor compound. Anti-inflammatory activity by downregulating COX-2 and iNOS expression via modulation of NF-kappaB activation. Antinociceptive. Probe to study chemokine receptors functioning. Potential therapeutic agent for osteoporosis and cancer-associated bone loss. Anti diabetic nephropathy

Formula: C₁₅H₂₂O
Storage Temperature: Freezer
MDL Number: MFCD03700769
CAS Number: 471-05-6

Order Now

Specification Test Results

Source/Host Isolated from the underground parts of Curcuma zerumbet.
Purity 99% (HPLC)
Appearance/Color Solid white crystals or powder.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR