Human Recombinant TNFR1 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0074-C050
AG-40B-0074-3050
102515-538EA
249.23
USD
102515-538
102515-540
Human Recombinant TNFR1 (from HEK293 Cells)
Proteins and Peptides
TNF-R1 is a receptor for TNF-alpha and homotrimeric lymphotoxin-alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis.