Human Recombinant Netrin-1 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0040-C010 AG-40B-0040-3010 AG-40B-0040-C100
102515-494EA 346.15 USD
102515-494 102515-496 102980-126
Human Recombinant Netrin-1 (from HEK293 Cells)
Proteins and Peptides
Netrin-1 controls guidance of CNS commissural axons and peripheral motor axons. Its association with either DCC or some UNC5 receptors will lead to axon attraction or repulsion, respectively. It also serve as a survival factor via its association with its receptors which prevent the initiation of apoptosis. Netrin-1 is also Involved in tumorigenesis by regulating apoptosis. Netrin-1 promotes atherosclerosis by retaining macrophages in the artery wall. Netrin-1 also governs induced pluripotent stem cell (iPS) formation and improves reprogramming efficiency of human and mouse somatic cells by limiting apoptosis mediated by Netrin-1 receptors DCC or UNC5b.