Human Recombinant IL-1 (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0024-C050 AG-40B-0024-3050
102515-454EA 360 USD
102515-454 102515-456
Human Recombinant IL-1 (from HEK293 cells)
Proteins and Peptides
IL-1RI is the receptor for interleukin-1alpha (IL-1A), beta (IL-1B) and interleukin-1 receptor antagonist protein (IL-1RA). Binding to the agonist leads to the activation of NF-kappaB. Signaling involves formation of a ternary complex containing IL1RAP, leading the activation of TOLLIP, MYD88 and IRAK1 or IRAK2.