Human Recombinant IL1B (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40B-0023-C010 AG-40B-0023-3010
102515-450EA 249.23 USD
102515-450 102515-452
Human Recombinant IL1B (from E. coli)
Proteins and Peptides
IL-1beta is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1beta belongs to the IL-1 family of proteins that are involved in the inflammatory response. IL-1beta in most situations requires the inflammasome complex to be cleaved and secreted.