Mouse Recombinant GITR (from HEK293 cells)
Supplier: ADIPOGEN CORP MS
AG-40B-0002-C050 AG-40B-0002-3050
102515-404EA 401.54 USD
102515-404 102515-406
Mouse Recombinant GITR (from HEK293 cells)
Proteins and Peptides
GITR (Glucocorticoid-induced TNF receptor) is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T cell receptor-mediated cell death. GITR serves as a negative regulator for NK cell activation. It mediates NF-kappaB activation via the TRAF2/NIK pathway.