Recombinant Listeria Monocytogenes P6 (from E. coli)
Supplier: ADIPOGEN CORP MS
AG-40A-0192-C010 AG-40A-0192-C050
102515-380EA 235.38 USD
102515-380 102515-382
Recombinant Listeria Monocytogenes P6 (from E. coli)
Proteins and Peptides
Listeria monocytogenes is a intracellular bacterium that is the causative agent of Listeriosis. It is one of the most virulent foodborne pathogens. Listeria species are known to secrete a 60kDa protein collectively termed p60, which is encoded by the "iap" (invasion-associated-protein) gene and secreted in large quantities into the growth media. p60 is a highly immunogenic murein hydrolase that is essential for cell division.