Human Recombinant DLL1 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0073-C010 AG-40A-0073-C050
102514-936EA 221.54 USD
102514-936 102514-938
Human Recombinant DLL1 (from HEK293 Cells)
Proteins and Peptides
The Notch ligand delta-like protein 1 (DLL1) is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T cell/NK-cell precursor.