Human Recombinant MFAP4 (from HEK293 Cells)
Supplier: ADIPOGEN CORP MS
AG-40A-0072-C010 AG-40A-0072-C050
102514-932EA 249.23 USD
102514-932 102514-934
Human Recombinant MFAP4 (from HEK293 Cells)
Proteins and Peptides
Microfibril-associated protein 4 (MFAP4) was initially identified as a gene commonly deleted in the contiguous Smith-Magenis syndrome (SMS) gene. MFAP4 was identified as growth factor to support expansion of hematopoietic stem cells in culture. It is involved in calcium-dependent cell adhesion and intercellular interactions.