Pierce™ TMT™ Elution Buffer
Supplier: LIFE TECHNOLOGIES MS
90104
PI90104EA 131.69 USD
PI90104
Pierce™ TMT™ Elution Buffer
Buffers Assay Buffers
Thermo Scientific TMT Elution Buffer is for use with Thermo Scientific Immobilized Anti-TMT Resin to enrich samples for peptides that have been chemically labeled with Tandem Mass Tag™ (TMT™) Reagents.
  • For use with a mass spectrometer
  • Designed to enrich chemically labeled samples for peptides
  • Helps provide higher specific enrichment compared to acidic or basic elution buffers
Immobilized Anti-TMT Resin and TMT Elution Buffer are effective for reducing sample complexity, improving dynamic range and studying cysteine modifications such as S-nitrosylation, oxidation and disulfide bonds of protein samples labeled with iodoTMT Reagents. This approach of selective thiol labeling and affinity enrichment is similar to isotope-coded affinity tags (ICAT™ Method) but allows for higher multiplex quantitation.
Caution: For Research Use Only. Not for use in diagnostic procedures.