Pierce™ Primary Neuron Isolation Kit, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
88280
PI88280EA 706.46 USD
PI88280
Pierce™ Primary Neuron Isolation Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Pierce™ Primary Neuron Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary neurons from embryonic cortical and hippocampal tissue from mouse and rat.
  • Procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Provides a 2-fold increase in yield compared to do-it-yourself methods
  • Greater than 95% of isolated cells remain viable in culture from Day 1 to Week 4
  • Cultured neurons contain processes indicative of complete differentiation and functionality
The Primary Neuron Isolation Kit has been designed to obtain high viability and purity of primary embryonic neurons using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains tissue-specific dissociation enzymes, media formulated specifically for neuronal cell culture, and cell culture supplements that promote the isolation and growth of functional neurons. The validated protocol and reagents produce highly pure primary neurons for either short- or long-term culture, and the kit is designed for use by both experienced and non-experienced users.
Ordering information: Upon receipt, store Product 8828X at 4?C and Product 8828Y at -20?C. Product 8828X is shipped with an ice pack. Product 8828Y is shipped on dry ice.
Delivery information: Kit includes Neuronal Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), Neuronal Culture Medium, Neuronal Culture Media Supplement, Glutamine Supplement, and Neuronal Growth Supplement. Also required (but not supplied) are fresh embryonic mouse or rat tissue sample, heat-inactivated foetal bovine serum (FBS), and poly-D-lysine treated culture dishes.
Caution: For Research Use Only. Not for use in diagnostic procedures.