Pierce™ 1-Step Transfer Buffer, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
84731X5
PI84731X5PK 444.23 USD
PI84731X5
Pierce™ 1-Step Transfer Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Transfer Buffers
Thermo Scientific Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
Features of 1-Step Transfer Buffer:
Fast—high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
Ready to use—supplied as a 1X solution that is stable and ready to use at room temperature
Optimized—designed for seamless and reliable performance with the Pierce G2 Fast Blotter
Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217)
Economical—excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices
When used with the
Fast—high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems