Pierce™ Substrate for Western Blotting, ECL Plus, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
80196
PI80196EA 505.2 USD
PI80196
Pierce™ Substrate for Western Blotting, ECL Plus, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The Pierce™ ECL Plus Western Blotting Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.
  • Easy to use, high quality and performance
  • Higher sensitivity - detect targets down to the low-picogram level
  • Longer signal duration - sustained light output for as long as 5 hours
  • More imaging options - X-ray, CCD or laser-based imagers
Pierce™ ECL 2 Substrate generates acridinium esters when it reacts with HRP
Pierce™ ECL 2 substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. A broad dynamic range and excellent sensitivity combined with a long-lasting signal make Pierce ECL 2 Substrate a desirable Western blotting substrate.
As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers.
Delivery information: Kit includes proprietary acridan and peroxide solutions.