Pierce™ Immobilised TCEP Disulfide Reducing Gel, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
77712
PI77712EA 274.46 USD
PI77712
Pierce™ Immobilised TCEP Disulfide Reducing Gel, Thermo Scientific
Protein Purification Protein Purification Reagents
Immobilised TCEP Disulfide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample from the reducing agent.
  • Odourless and virtually no loss of activity when stored refrigerated
  • Reduces peptides and proteins over a broad range of pH, salt or detergent concentrations, and temperature conditions
  • Resin slurry provides flexibility for use in spin tube, column or batch modes
  • No soluble reductant removal saves time and conserves sample
  • Cleaves both natural disulfide bonds and those incorporated by crosslinking