Pierce Imject™ Maleimide Conjugation Buffer, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
77164
PI77164EA 125.77 USD
PI77164
Pierce Imject™ Maleimide Conjugation Buffer, Thermo Scientific
Buffers Antibody Purification Buffers
Imject™ maleimide conjugation buffer (30 mL) is sufficient for dissolving up to 60 mg of sulfhydryl-peptide for maleimide conjugation with carrier proteins.
Formulation: 0.083 M sodium phosphate, 0.1 M EDTA, 0.9 M NaCl, 0.1 M sorbitol and 0.02% sodium azide.
Caution: For research use only. Not for use in diagnostic procedures.