Pierce Imject™ Purification Buffer Salts, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
77159
PI77159EA 92.69 USD
PI77159
Pierce Imject™ Purification Buffer Salts, Thermo Scientific
Buffers Antibody Purification Buffers
Imject™ purification buffer salts is a dry powder for preparing 60 mL of solution for buffer-exchange of immunogens following conjugation.
Caution: For research use only. Not for use in diagnostic procedures.