5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] mixture of isomers fluorescent dye, Pierce™

Supplier: LIFE TECHNOLOGIES MS

46406
PI46406EA 224 USD
PI46406
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] mixture of isomers fluorescent dye, Pierce™
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]
Formula: C₂₉H₂₅N₃O₇
MDL Number: MFCD00077324
CAS Number: 150408-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR