Pierce 1-Step™ NBT/BCIP plus Suppressor Substrate Solution, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
34070
PI34070EA 184.62 USD
PI34070
Pierce 1-Step™ NBT/BCIP plus Suppressor Substrate Solution, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The 1-Step™ NBT and BCIP substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.
  • Sensitive with low background and minimial assay-to-assay variability
  • Ready-to-use single component
The combination of NBT and BCIP yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, an enzyme conjugate for antibody probes. Contains 1 mM levamisole to inhibit endogenous phosphatase activity, while yielding an intense dark purple precipitate in the presence of calf intestinal phosphatase. Used for immunoblotting, immunohistochemistry, and in situ hybridization applications.