Pierce 1-Step™ TMB-Blotting Substrate Solution, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
34018
PI34018EA 226.26 USD
PI34018
Pierce 1-Step™ TMB-Blotting Substrate Solution, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The 1-Step™ TMB-blotting solution is a blue TMB peroxidase (HRP) substrate that is specially formulated for chromogenic detection in Western blot and IHC experiments.
  • Ready-to-use solution
  • Highly sensitive and generates a blue-purple precipitate