Sodium dodecyl sulfate (SDS), Pierce™

Supplier: LIFE TECHNOLOGIES MS
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

28364 28365
PI28364EA 132.92 USD
PI28364 PI28365
Sodium dodecyl sulfate (SDS), Pierce™
Sodium dodecyl sulphate (SDS)

Thermo Scientific Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).


  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis
  • Common component of cell lysis buffers


Properties of SDS (values for pure C12):
Detergent Class: Ionic (anionic)
Aggregation Number: 62
Micelle Molecular Weight: 18,000 g
Critical Micelle Concentration (CMC): 6 to 8 mM (0.1728 to 0.2304%, w/v)
Cloud Point: >100°C
Dialyzable: No


Ordering information: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR