Pierce™ 10X Tris-Glycine Buffer, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
28363
PI28363EA 113.54 USD
PI28363
Pierce™ 10X Tris-Glycine Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Transfer Buffers
These space-saving stock solutions are ideal for quick preparation of standard Tris-glycine (pH 8.5) transfer buffer which is used for Western blotting.
  • Easy to use
  • Eliminates the possibility of powder remaining in a packet
  • Eliminates time spent waiting for powder to dissolve
Required:
Diluted 10-fold in water or 20% methanol, the solution yields 0.025M Tris, 0.192M glycine, pH 8.5
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Caution: For Research Use Only. Not for use in diagnostic procedures.