Pierce™ Western Blotting Kits, Far-Western Blot Kit for Biotinylated Proteins, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
23500
PI23500EA 851.69 USD
PI23500
Pierce™ Western Blotting Kits, Far-Western Blot Kit for Biotinylated Proteins, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The Pierce™ Far-Western Blot Kit for Biotinylated Proteins provide a nonisotopic approach to far-Western analysis that can be performed either directly in the gel or on the membrane
  • Reliable and sensitive detection method offers a practical and safe alternative to radiolabeling the bait protein
  • Primary antibody-free detection - uses a biotinylated protein as the probe, eliminating the need for primary antibodies or even to have prior knowledge of target-protein identities
  • Compatible with both SDS-PAGE and native gels
  • Biotin/streptavidin-HRP systems demonstrate less nonspecific binding compared to antibodies
  • Gels can be used for total protein staining after the in-gel chemiluminescent detection step, eliminating the need to run two gels
On-membrane detection offers greater sensitivity whereas in-gel detection is faster and prevents problems associated with incomplete or inefficient transfer.
Delivery information: Kit includes Streptavidin-HRP Conjugate (0.1 mg), 10X Dilution Buffer (50 mL), PBS Pack (makes 500 mL, 17 pack), Luminol Enhancer (55 mL), Stable Peroxide Buffer (55 mL), Surfact-Amps 20 Detergent Solution (6x10 mL) and Pre-cut Cellophane (10 sheets).