Pierce™ Streptavidin, Hydrazide-Activated, Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
21120
PI21120EA 254.77 USD
PI21120
Pierce™ Streptavidin, Hydrazide-Activated, Thermo Scientific
Protein Purification Protein Purification Reagents
Pierce™ Hydrazide-activated strepavidin conjugate includes recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.