Cysteaminium chloride, Pierce™

Supplier: LIFE TECHNOLOGIES MS

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

20408
PI20408EA 289.23 USD
PI20408
Cysteaminium chloride, Pierce™
Cysteaminium chloride

Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₂H₇NS·HCl
MW: 113.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR