Pierce™ Firefly Signal Enhancer (100X), Thermo Scientific
Supplier: LIFE TECHNOLOGIES MS
16180
PI16180EA 60.91 USD
PI16180
Pierce™ Firefly Signal Enhancer (100X), Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
Thermo Scientific Pierce Firefly Signal Enhancer is designed to complement the Pierce Firefly Luciferase Flash and Glow Assay and is recommended when maximum sensitivity is desired.
  • Up to 5-fold increase in Firefly Flash signal intensity
  • Up to 2-fold increase in Firefly Glow signal intensity and duration
  • Convenient and easy-to-use 100X solution
Caution: For Research Use Only. Not for use in diagnostic procedures.