2-Propanol ≥99.5%, Cleanroom® LP for the electronics industry

Supplier: KMG
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

200440
KM431-200440CS 538.39 USD
KM431-200440
2-Propanol ≥99.5%, Cleanroom® LP for the electronics industry
2-Propanol

IPA is used to rinse silicon wafers and to leave a clean dry residue free surface usually after organic stripping solutions

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C3H8O) 99.5% Min.
Assay (H2O) 500 ppm
Color 10 APHA
Appearance Conforms
Acidity 0.200 μeq/g
Alkalinity 0.100 μeq/g
Residue After Evaporation 4,000 ppb
Solubility (in H2O) Conforms
Infrared Conforms
Chloride (Cl) 100 ppb
Phosphate (PO4) 300 ppb
Aluminum (Al) 50.0 ppb
Antimony (Sb) 5.0 ppb
Arsenic (As) 5.0 ppb
Barium (Ba) 100.0 ppb
Beryllium (Be) 100.0 ppb
Bismuth (Bi) 100.0 ppb
Boron (B) 10.0 ppb
Cadmium (Cd) 100.0 ppb
Calcium (Ca) 50.0 ppb
Chromium (Cr) 50.0 ppb
Cobalt (Co) 50.0 ppb
Copper (Cu) 10.0 ppb
Gallium (Ga) 50.0 ppb
Germanium (Ge) 50.0 ppb
Gold (Au) 50.0 ppb
Iron (Fe) 50.0 ppb
Lead (Pb) 50.0 ppb
Lithium (Li) 50.0 ppb
Magnesium (Mg) 50.0 ppb
Manganese (Mn) 15.0 ppb
Molybdenum (Mo) 100.0 ppb
Nickel (Ni) 10.0 ppb
Niobium (Nb) 100.0 ppb
Potassium (K) 75.0 ppb
Silicon (Si) 100.0 ppb
Silver (Ag) 50.0 ppb
Sodium (Na) 75.0 ppb
Strontium (Sr) 100.0 ppb
Tantalum (Ta) 100.0 ppb
Thallium (Tl) 100.0 ppb
Tin (Sn) 50.0 ppb
Titanium (Ti) 50.0 ppb
Vanadium (V) 100.0 ppb
Zinc (Zn) 50.0 ppb
Zirconium (Zr) 100.0 ppb
0.2μ Particle Count 1,000 par/ml
0.3μ Particle Count 500 par/ml
0.5μ Particle Count 50 par/ml
1.0μ Particle Count 8 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR