Acetone ≥99.5%, Cleanroom® LP for the electronics industry

Supplier: KMG
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

200406
KM402-200406CS 750.34 USD
KM402-200406
Acetone ≥99.5%, Cleanroom® LP for the electronics industry
Acetone

Acetone can be used for drying laboratory glassware, removing greasy or oily contaminants from work benches, or as a general photoresist stripper or edgebead remover.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067

Order Now

Specification Test Results

Assay (Acetone) 99.5% min
Assay (H2O) 0,5%
Assay (C3H8O) 0,05%
Assay (CH3OH) 0,05%
Color 10 APHA
Appearance Conforms
Acidity 0.300 μeq/g
Alkalinity 0.300 μeq/g
Aldehydes 20 ppm
Residue After Evaporation 5000 ppb
Solubility (in H2O) Conforms
Substance Reducing KMnO4 Conforms
Infared Conforms
Chloride (Cl) 200 ppb
Phosphate (PO4) 50 ppb
Sulfate (SO4) 200 ppb
Antimony (Sb) 5 ppb
Arsenic (As) 5 ppb
Barium (Ba) 50 ppb
Beryllium (Be) 50 ppb
Bismuth (Bi) 50 ppb
Boron (B) 50 ppb
Cadmium (Cd) 50 ppb
Calcium (Ca) 50 ppb
Chromium (Cr) 50 ppb
Cobalt (Co) 10 ppb
Copper (Cu) 10 ppb
Gallium (Ga) 50 ppb
Germanium (Ge) 50 ppb
Gold (Au) 50 ppb
Iron (Fe) 50 ppb
Lead (Pb) 50 ppb
Lithium (Li) 50 ppb
Magnesium (Mg) 50 ppb
Manganese (Mn) 50 ppb
Molybdenum (Mo) 50 ppb
Nickel (Ni) 10 ppb
Niobium (Nb) 50 ppb
Potassium (K) 50 ppb
Silicon (Si) 50 ppb
Silver (Ag) 50 ppb
Strontium (Sr) 50 ppb
Tantalum (Ta) 50 ppb
Thallium (Tl) 50 ppb
Tin (Sn) 50 ppb
Titanium (Ti) 50 ppb
Vanadium (V) 50 ppb
Zinc (Zn) 50 ppb
Zirconium (Zr) 50 ppb
1.0μ Particle Count 10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR