DSP (Dithiobis(succinimidyl propionate)) ≥94% (by HPLC), water soluble, Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: DTSSP, 3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), DSP Crosslinker, Lomant’s reagent, 3,3'-Dithiodipropionic acid bis(N-succinimidyl Ester), Di(N-succinimidyl)-3,3'-dithiodipropionate, Sulfo-DSP, DTSP, 3,3'-Dithiodipropionic acid di(N-succinimidyl Ester)

M-Clarity™ quality level = MQ100

322133-100MG
EM322133-100MGEA 322.14 USD
EM322133-100MG
DSP (Dithiobis(succinimidyl propionate)) ≥94% (by HPLC), water soluble, Calbiochem®, Sigma-Aldrich®
DSP (Dithiobis(succinimidyl propionate))

Bifunctional cross-linker. The disulfide bond can be cleaved with 1% NaBH4, 50 mM DTT, or 100 mM β-mercaptoethanol. Has been used to immobilize proteins on supports containing amino groups.

Formula: C₁₄H₁₆N₂O₈S₂
MW: 404.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042045
CAS Number: 57757-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR