MC1061 F-Competent Cells, Biosearch Technologies
Supplier: Lucigen
60514-1 60514-2
89260-194EA 522.39 USD
89260-194 89260-196
MC1061 F-Competent Cells, Biosearch Technologies
Competent Cells
MC1061 F- Electrocompetent Cells
An amber suppressor strain, identical to SS320 except that it lacks the F′episome. Highly efficient (≥4 × 1010 cfu/µg) electrocompetent cells for general cloning or Phage Display. Note that as an F- strain, it cannot be used for re-infection by filamentous phage