DSP (Dithiobis(succinimidyl propionate))

Supplier: G-Biosciences

Synonyms: DTSSP, 3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), DSP Crosslinker, Lomant’s reagent, 3,3'-Dithiodipropionic acid bis(N-succinimidyl Ester), Di(N-succinimidyl)-3,3'-dithiodipropionate, Sulfo-DSP, DTSP, 3,3'-Dithiodipropionic acid di(N-succinimidyl Ester)

BC07
82022-782EA 509.69 USD
82022-782
DSP (Dithiobis(succinimidyl propionate))
DSP (Dithiobis(succinimidyl propionate))

Dithiobis(succinimidyl propionate) Homobifunctional Cross Linker.

Formula: C₁₄H₁₆N₂O₈S₂
MW: 404.41 g/mol
MDL Number: MFCD00042045
CAS Number: 57757-57-0

Order Now

Specification Test Results

Spacer Arm (Å) 12
Reactive Toward Primary Amines
Membrane Permeable 1
Water Soluble 0
Cleavable/ Reversible Reducing Agents (Thiols)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR