PCP-d5 (hydrochloride) (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
ISO60200-1
76544-728EA 137 USD
76544-728
PCP-d5 (hydrochloride) (CRM), Cayman Chemical
Standards Organic Standards
A Certified Reference Material
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard