Codeine (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
ISO60140-1
76544-626EA 25 USD
76544-626
Codeine (CRM), Cayman Chemical
Standards Organic Standards
A Certified Reference Material
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43a2510e5831ed6325862aa1b91ba804760e0f01&vid=43a2510e5831ed6325862aa1b91ba804760e0f01&prid=web&clid=SAPDAM