Adinazolam (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
30186-1
76544-522EA 126 USD
76544-522
Adinazolam (CRM), Cayman Chemical
Standards Organic Standards
A Certified Reference Material
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08b08de7908931a99ec5caf89b6b04ed116728f2&vid=08b08de7908931a99ec5caf89b6b04ed116728f2&prid=web&clid=SAPDAM