Mycotoxin Mixture 5 (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
28151-1
76544-494EA
461
USD
76544-494
Mycotoxin Mixture 5 (CRM), Cayman Chemical
Standards
Organic Standards
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)