Phytocannabinoid Mixture 3 (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
23251-1
76544-324EA 145 USD
76544-324
Phytocannabinoid Mixture 3 (CRM), Cayman Chemical
Standards Organic Standards
A mixture of phytocannabinoids in a methanol solution intended for HPLC-UV or LC- or GC-MS
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard