Zaleplon (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
20411-1
76544-194EA 96 USD
76544-194
Zaleplon (CRM), Cayman Chemical
Standards Organic Standards
A Certified Reference Material
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9327daaf56eeab90ca833e60ed9f6b639114b5bd&vid=9327daaf56eeab90ca833e60ed9f6b639114b5bd&prid=web&clid=SAPDAM