Nimetazepam (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
19206-1
76544-064EA 30 USD
76544-064
Nimetazepam (CRM), Cayman Chemical
Standards Organic Standards
A Certified Reference Material
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard