N-Desmethylclobazam (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
18888-100
76544-034EA 141 USD
76544-034
N-Desmethylclobazam (CRM), Cayman Chemical
Standards Organic Standards
A major active metabolite of clobazam that positively modulates GABAA receptors (EC50s range from 98 to 151 µM, depending on subunit configuration)
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard